trans-3-Hydroxycyclobutanecarbonitrile

Lieferant: Apollo Scientific

OR304377-1G
APOSOR304377-1GEA 1160 EUR
APOSOR304377-1G
trans-3-Hydroxycyclobutanecarbonitrile
trans-3-Hydroxycyclobutanecarbonitrile
Formel: C₅H₇NO
Molecular Weight: 97.115 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30802788
CAS-Nummer: 88315-80-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR