[cis-3-Methoxycyclobutyl]methanamine hydrochloride
Lieferant: Apollo Scientific
OR304379-1G
APOSOR304379-1GEA
1420
EUR
APOSOR304379-1G
[cis-3-Methoxycyclobutyl]methanamine hydrochloride
[cis-3-Methoxycyclobutyl]methanamine hydrochloride
Formel:
C₆H₁₃NO·ClH Molecular Weight: 151,634 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30488043 CAS-Nummer: 2044706-13-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...