trans-3-Methoxycyclobutane-1-methamine hydrochloride

Lieferant: Apollo Scientific

OR304380-1G
APOSOR304380-1GEA 1710 EUR
APOSOR304380-1G
trans-3-Methoxycyclobutane-1-methamine hydrochloride
trans-3-Methoxycyclobutane-1-methamine hydrochloride
Formel: C₆H₁₃NO·ClH
Molecular Weight: 151,634 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30734068
CAS-Nummer: 2094029-36-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR