3-Ethoxy-2,2-dimethylcyclobutan-1-amine
Lieferant: Apollo Scientific
OR304381-1G
APOSOR304381-1GEA
833
EUR
APOSOR304381-1G
3-Ethoxy-2,2-dimethylcyclobutan-1-amine
3-Ethoxy-2,2-dimethylcyclobutan-1-amine
Formel:
C₈H₁₇NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD19982396 CAS-Nummer: 1333759-16-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...