1-(Prop-2-en-1-yl)cyclobutane-1-carboxylic acid
Lieferant: Apollo Scientific
OR304382-1G
OR304382-5G
APOSOR304382-1GEA
589
EUR
APOSOR304382-1G
APOSOR304382-5G
1-(Prop-2-en-1-yl)cyclobutane-1-carboxylic acid
1-(Prop-2-en-1-yl)cyclobutane-1-carboxylic acid
Formel:
C₈H₁₂O₂ Molecular Weight: 140.18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19229282 CAS-Nummer: 1193397-42-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...