cis-2-Aminocyclobutan-1-ol hydrochloride

Lieferant: Apollo Scientific

OR304384-1G
APOSOR304384-1GEA 1010 EUR
APOSOR304384-1G
cis-2-Aminocyclobutan-1-ol hydrochloride
cis-2-Aminocyclobutan-1-ol hydrochloride
Formel: C₄H₉NO·ClH
Molecular Weight: 123,581 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26127763
CAS-Nummer: 2070860-49-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR