trans-3-(2-Hydroxypropan-2-yl)cyclobutane-1-carboxylic acid

Lieferant: Apollo Scientific

OR304385-500MG
APOSOR304385-500MGEA 1830 EUR
APOSOR304385-500MG
trans-3-(2-Hydroxypropan-2-yl)cyclobutane-1-carboxylic acid
trans-3-(2-Hydroxypropan-2-yl)cyclobutane-1-carboxylic acid
Formel: C₈H₁₄O₃
Molecular Weight: 158,195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30803329
CAS-Nummer: 1940136-70-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR