2-[trans-3-Aminocyclobutyl]propan-2-ol hydrochloride

Lieferant: Apollo Scientific

OR304386-1G
APOSOR304386-1GEA 1490 EUR
APOSOR304386-1G
2-[trans-3-Aminocyclobutyl]propan-2-ol hydrochloride
2-[trans-3-Aminocyclobutyl]propan-2-ol hydrochloride
Formel: C₇H₁₅NO·ClH
Molecular Weight: 165,661 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30803654
CAS-Nummer: 2058249-61-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR