1-(4-Methylpyridazin-3-yl)methanamine hydrochloride

Lieferant: Apollo Scientific

OR304391-1G
APOSOR304391-1GEA 2050 EUR
APOSOR304391-1G
1-(4-Methylpyridazin-3-yl)methanamine hydrochloride
1-(4-Methylpyridazin-3-yl)methanamine hydrochloride
Formel: C₆H₉N₃·ClH
Molecular Weight: 159,617 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31567447
CAS-Nummer: 2173991-77-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR