5-Methylpyridazine-4-carbonitrile
Lieferant: Apollo Scientific
OR304393-1G
OR304393-250MG
OR304393-500MG
APOSOR304393-1GEA
1270
EUR
APOSOR304393-1G
APOSOR304393-250MG
APOSOR304393-500MG
5-Methylpyridazine-4-carbonitrile
5-Methylpyridazine-4-carbonitrile
Formel:
C₆H₅N₃ Molecular Weight: 119.124 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30471638 CAS-Nummer: 2007921-31-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...