4,7-Diazaspiro[2.5]octan-5-one
Lieferant: Apollo Scientific
OR304394-1G
APOSOR304394-1GEA
2220
EUR
APOSOR304394-1G
4,7-Diazaspiro[2.5]octan-5-one
4,7-Diazaspiro[2.5]octan-5-one
Formel:
C₆H₁₀N₂O Molecular Weight: 126,156 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20526179 CAS-Nummer: 1200114-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...