5,8-Diazaspiro[3.5]nonan-6-one

Lieferant: Apollo Scientific

OR304395-1G
APOSOR304395-1GEA 2890 EUR
APOSOR304395-1G
5,8-Diazaspiro[3.5]nonan-6-one
5,8-Diazaspiro[3.5]nonan-6-one
Formel: C₇H₁₂N₂O
Molecular Weight: 140,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31567418
CAS-Nummer: 1419209-31-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR