6-Methoxy-2-azaspiro[3.3]heptane hydrochloride

Lieferant: Apollo Scientific

OR304397-1G
APOSOR304397-1GEA 307 EUR
APOSOR304397-1G
6-Methoxy-2-azaspiro[3.3]heptane hydrochloride
6-Methoxy-2-azaspiro[3.3]heptane hydrochloride
Formel: C₇H₁₃NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD28501414
CAS-Nummer: 1638761-19-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR