5-Oxaspiro[2.4]heptan-6-one

Lieferant: Apollo Scientific

OR304399-1G
APOSOR304399-1GEA 1150 EUR
APOSOR304399-1G
5-Oxaspiro[2.4]heptan-6-one
5-Oxaspiro[2.4]heptan-6-one
Formel: C₆H₈O₂
Molecular Weight: 112,127 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17014664
CAS-Nummer: 33867-36-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR