7-Amino-2-azaspiro[4.4]nonan-3-one hydrochloride

Lieferant: Apollo Scientific

OR304401-1G
APOSOR304401-1GEA 3280 EUR
APOSOR304401-1G
7-Amino-2-azaspiro[4.4]nonan-3-one hydrochloride
7-Amino-2-azaspiro[4.4]nonan-3-one hydrochloride
Formel: C₈H₁₄N₂O·ClH
Molecular Weight: 190,671 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31617948
CAS-Nummer: 2173991-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR