2,9-Diazaspiro[5.5]undecane dihydrochloride

Lieferant: Apollo Scientific

OR304403-1G
APOSOR304403-1GEA 0 EUR
APOSOR304403-1G
2,9-Diazaspiro[5.5]undecane dihydrochloride
2,9-Diazaspiro[5.5]undecane dihydrochloride
Formel: C₉H₁₈N₂·2ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD10700111
CAS-Nummer: 2227206-72-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR