7-Benzyl-1,7-diazaspiro[3.5]nonane

Lieferant: Apollo Scientific

OR304406-1G
APOSOR304406-1GEA 1490 EUR
APOSOR304406-1G
7-Benzyl-1,7-diazaspiro[3.5]nonane
7-Benzyl-1,7-diazaspiro[3.5]nonane
Formel: C₁₄H₂₀N₂
Molecular Weight: 216,322 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24384637
CAS-Nummer: 1420897-28-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR