6-Methyl-2-azaspiro[3.3]heptane hydrochloride

Lieferant: Apollo Scientific

Synonyme: 6-methyl-2-azaspiro[3.3]heptane hydrochloride

OR304407-1G
APOSOR304407-1GEA 811 EUR
APOSOR304407-1G
6-Methyl-2-azaspiro[3.3]heptane hydrochloride
6-Methyl-2-azaspiro[3.3]heptane hydrochloride
Formel: C₇H₁₃N·ClH
Molecular Weight: 147,646 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30829398
CAS-Nummer: 2089649-42-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR