Spiro[3.4]octan-2-one

Lieferant: Apollo Scientific

OR304408-1G
APOSOR304408-1GEA 911 EUR
APOSOR304408-1G
Spiro[3.4]octan-2-one
Spiro[3.4]octan-2-one
Formel: C₈H₁₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD19441113
CAS-Nummer: 41463-77-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR