2-(Isopropylsulfonyl)anilin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Isopropylsulphonyl)aniline, 2-Aminophenyl Isopropyl sulphone
OR30441-500MG
APOSOR30441-500MGEA
21.9
EUR
APOSOR30441-500MG
2-(Isopropylsulfonyl)anilin 98%
1-Amino-2-(isopropylsulphonyl)benzene
Formel:
C₉H₁₃NO₂S Molecular Weight: 199,2700 g/mol Schmelzpunkt: 83 - 84 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08445603 CAS-Nummer: 76697-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...