7-Methyl-2,7-diazaspiro[4.5]decane dihydrochloride

Lieferant: Apollo Scientific

OR304410-1G
APOSOR304410-1GEA 1150 EUR
APOSOR304410-1G
7-Methyl-2,7-diazaspiro[4.5]decane dihydrochloride
7-Methyl-2,7-diazaspiro[4.5]decane dihydrochloride
Formel: C₉H₁₈N₂·2ClH
Molecular Weight: 227,175 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26407167
CAS-Nummer: 1086395-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR