1,6-Diazaspiro[3.5]nonan-2-one

Lieferant: Apollo Scientific

OR304414-1G
APOSOR304414-1GEA 1150 EUR
APOSOR304414-1G
1,6-Diazaspiro[3.5]nonan-2-one
1,6-Diazaspiro[3.5]nonan-2-one
Formel: C₇H₁₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD26858689
CAS-Nummer: 1567085-15-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR