5-Oxaspiro[3.5]nonan-8-ol
Lieferant: Apollo Scientific
OR304415-1G
APOSOR304415-1GEA
1040
EUR
APOSOR304415-1G
5-Oxaspiro[3.5]nonan-8-ol
5-Oxaspiro[3.5]nonan-8-ol
Formel:
C₈H₁₄O₂ Molecular Weight: 142,196 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22070788 CAS-Nummer: 1368181-23-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...