5-Oxaspiro[3.5]nonan-8-one
Lieferant: Apollo Scientific
OR304416-1G
APOSOR304416-1GEA
1040
EUR
APOSOR304416-1G
5-Oxaspiro[3.5]nonan-8-one
5-Oxaspiro[3.5]nonan-8-one
Formel:
C₈H₁₂O₂ Molecular Weight: 140,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22070990 CAS-Nummer: 1367945-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...