7-Oxospiro[3.5]nonane-2-carboxylic acid

Lieferant: Apollo Scientific

OR304417-1G
APOSOR304417-1GEA 1860 EUR
APOSOR304417-1G
7-Oxospiro[3.5]nonane-2-carboxylic acid
7-Oxospiro[3.5]nonane-2-carboxylic acid
Formel: C₁₀H₁₄O₃
Molecular Weight: 182,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24857339
CAS-Nummer: 1440962-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR