Spiro[3.5]nonan-1-ol
Lieferant: Apollo Scientific
OR304418-1G
APOSOR304418-1GEA
1150
EUR
APOSOR304418-1G
Spiro[3.5]nonan-1-ol
Spiro[3.5]nonan-1-ol
Formel:
C₉H₁₆O Lagertemperatur: Raumtemperatur |
MDL:
MFCD26407277 CAS-Nummer: 60211-19-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...