Spiro[3.5]nonan-1-one

Lieferant: Apollo Scientific

OR304419-1G
APOSOR304419-1GEA 923 EUR
APOSOR304419-1G
Spiro[3.5]nonan-1-one
Spiro[3.5]nonan-1-one
Formel: C₉H₁₄O
Molecular Weight: 138,207 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24386783
CAS-Nummer: 29800-45-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR