Spiro[2.3]hexan-5-one

Lieferant: Apollo Scientific

OR304420-1G
APOSOR304420-1GEA 359 EUR
APOSOR304420-1G
Spiro[2.3]hexan-5-one
Spiro[2.3]hexan-5-one
Formel: C₆H₈O
Molecular Weight: 96,127 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23884029
CAS-Nummer: 20061-22-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR