2,7-Diazaspiro[3.5]nonane dihydrochloride

Lieferant: Apollo Scientific

OR304422-1G
APOSOR304422-1GEA 923 EUR
APOSOR304422-1G
2,7-Diazaspiro[3.5]nonane dihydrochloride
2,7-Diazaspiro[3.5]nonane dihydrochloride
Formel: C₇H₁₄N₂·2ClH
Molecular Weight: 199,121 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20261446
CAS-Nummer: 1334499-85-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR