7-Oxa-1-azaspiro[3.5]nonane

Lieferant: Apollo Scientific

OR304425-1G
APOSOR304425-1GEA 1560 EUR
APOSOR304425-1G
7-Oxa-1-azaspiro[3.5]nonane
7-Oxa-1-azaspiro[3.5]nonane
Formel: C₇H₁₃NO
Molecular Weight: 127,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14581251
CAS-Nummer: 106404-52-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR