6-Oxa-1-azaspiro[3.5]nonane

Lieferant: Apollo Scientific

OR304426-1G
APOSOR304426-1GEA 3110 EUR
APOSOR304426-1G
6-Oxa-1-azaspiro[3.5]nonane
6-Oxa-1-azaspiro[3.5]nonane
Formel: C₇H₁₃NO
Molecular Weight: 127,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14581253
CAS-Nummer: 1214875-46-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR