4-Oxa-1,9-diazaspiro[5.5]undecane dihydrochloride

Lieferant: Apollo Scientific

OR304428-250MG OR304428-500MG
APOSOR304428-250MGEA 0 EUR
APOSOR304428-250MG APOSOR304428-500MG
4-Oxa-1,9-diazaspiro[5.5]undecane dihydrochloride
4-Oxa-1,9-diazaspiro[5.5]undecane dihydrochloride
Formel: C₈H₁₆N₂O·2ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD26096846
CAS-Nummer: 1439897-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR