3-lambda-Thia-9-azaspiro[5.5]undecane-3,3-dione hydrochloride

Lieferant: Apollo Scientific

OR304434-1G
APOSOR304434-1GEA 1710 EUR
APOSOR304434-1G
3-lambda-Thia-9-azaspiro[5.5]undecane-3,3-dione hydrochloride
3-lambda-Thia-9-azaspiro[5.5]undecane-3,3-dione hydrochloride
Formel: C₉H₁₇NO₂S·ClH
Molecular Weight: 239,763 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28954306
CAS-Nummer: 1803582-31-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR