{Spiro[2.5]octan-5-yl}methanol

Lieferant: Apollo Scientific

OR304437-1G
APOSOR304437-1GEA 1490 EUR
APOSOR304437-1G
{Spiro[2.5]octan-5-yl}methanol
{Spiro[2.5]octan-5-yl}methanol
Formel: C₉H₁₆O
Molecular Weight: 140,223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30803019
CAS-Nummer: 2002815-12-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR