(S)-5-Methyl-5-azaspiro[2.4]heptane-6-carboxylic acid
Lieferant: Apollo Scientific
OR304438-1G
OR304438-500MG
APOSOR304438-1GEA
1150
EUR
APOSOR304438-1G
APOSOR304438-500MG
(S)-5-Methyl-5-azaspiro[2.4]heptane-6-carboxylic acid
(S)-5-Methyl-5-azaspiro[2.4]heptane-6-carboxylic acid
Formel:
C₈H₁₃NO₂ Molecular Weight: 155.194 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30146460 CAS-Nummer: 1860033-47-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...