1-{2,7-Diazaspiro[3.5]nonan-7-yl}ethan-1-one hydrochloride

Lieferant: Apollo Scientific

OR304441-1G
APOSOR304441-1GEA 1830 EUR
APOSOR304441-1G
1-{2,7-Diazaspiro[3.5]nonan-7-yl}ethan-1-one hydrochloride
1-{2,7-Diazaspiro[3.5]nonan-7-yl}ethan-1-one hydrochloride
Formel: C₉H₁₆N₂O·ClH
Molecular Weight: 204,697 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31617951
CAS-Nummer: 2173992-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR