1-{2,7-Diazaspiro[3.5]nonan-2-yl}ethan-1-one

Lieferant: Apollo Scientific

OR304442-1G
APOSOR304442-1GEA 1640 EUR
APOSOR304442-1G
1-{2,7-Diazaspiro[3.5]nonan-2-yl}ethan-1-one
1-{2,7-Diazaspiro[3.5]nonan-2-yl}ethan-1-one
Formel: C₉H₁₆N₂O
Molecular Weight: 168,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28129054
CAS-Nummer: 1474026-47-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR