1-Methyl-1,6-diazaspiro[3.4]octane
Lieferant: Apollo Scientific
OR304443-1G
APOSOR304443-1GEA
2720
EUR
APOSOR304443-1G
1-Methyl-1,6-diazaspiro[3.4]octane
1-Methyl-1,6-diazaspiro[3.4]octane
Formel:
C₇H₁₄N₂ Molecular Weight: 126,199 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD12406513 CAS-Nummer: 1158749-83-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...