(3-Brom-4-methoxyphenyl)essigsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-4-methoxyphenylacetic acid, 2-(3-Bromo-4-methoxyphenyl)acetic acid

OR3045-25G
APOSOR3045-25GEA 87 EUR
APOSOR3045-25G
(3-Brom-4-methoxyphenyl)essigsäure
(3-Brom-4-methoxyphenyl)essigsäure
Formel: C₉H₉BrO₃
Molecular Weight: 245.07 g/mol
Schmelzpunkt: 115…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017538
CAS-Nummer: 774-81-2
UN: 3077
ADR: 9+EHS III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR