(5-Brom-2-methoxyphenyl)essigsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(5-Bromo-2-methoxyphenyl)acetic acid

OR3046-1G
APOSOR3046-1GEA 21.9 EUR
APOSOR3046-1G
(5-Brom-2-methoxyphenyl)essigsäure
(5-Brom-2-methoxyphenyl)essigsäure
Formel: C₉H₉BrO₃
Molecular Weight: 245.07 g/mol
Schmelzpunkt: 132…134 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016825
CAS-Nummer: 7017-48-3
UN: 3077
ADR: 9+EHS III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR