2,5-Dibromhydrochinon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,5-Dibromobenzene-1,4-diol

OR30490-10G OR30490-5G
APOSOR30490-10GEA 33.5 EUR
APOSOR30490-10G APOSOR30490-5G
2,5-Dibromhydrochinon 98%
2,5-Dibromhydrochinon
Formel: C₆H₄Br₂O₂
Molecular Weight: 267.9 g/mol
Schmelzpunkt: 193 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00192664
CAS-Nummer: 14753-51-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR