3-[(Diethoxyphosphoryl)oxy]-1,2,3-benzotriazin-4(3H)-on 98%
Lieferant: Apollo Scientific
Synonyme:
Diethyl (4-oxobenzo[d][1,2,3]triazin-3(4H)-yl) phosphate, DEPBT, Diethyl 3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl Phosphate
OR30492-100G
APOSOR30492-100GEA
119
EUR
APOSOR30492-100G
3-[(Diethoxyphosphoryl)oxy]-1,2,3-benzotriazin-4(3H)-on 98%
3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one
Formel:
C₁₁H₁₄N₃O₅P Molecular Weight: 299,22 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD01236967 CAS-Nummer: 165534-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...