Chinoxalin-2,3-dion 98%
Lieferant: Apollo Scientific
Synonyme:
2,3-Dihydroxyquinoxaline, Quinoxaline-2,3(1H,4H)-dione
OR30493-100G
APOSOR30493-100GEA
77
EUR
APOSOR30493-100G
Chinoxalin-2,3-dion 98%
Chinoxalin-2,3-dion
Formel:
C₈H₆N₂O₂ Molecular Weight: 162,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006723 CAS-Nummer: 15804-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...