4-Methoxybenzene-1,2-diol 95%

Lieferant: Apollo Scientific
Achtung

OR305004-1G OR305004-250MG OR305004-100MG
APOSOR305004-1GEA 386 EUR
APOSOR305004-1G APOSOR305004-250MG APOSOR305004-100MG
4-Methoxybenzene-1,2-diol 95%
4-Methoxybenzene-1,2-diol
Formel: C₇H₈O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18395726
CAS-Nummer: 3934-97-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR