Methyl 2-cyano-5-methoxybenzoate

Lieferant: Apollo Scientific

OR305013-250MG
APOSOR305013-250MGEA 71.2 EUR
APOSOR305013-250MG
Methyl 2-cyano-5-methoxybenzoate
Methyl 2-cyano-5-methoxybenzoate
Formel: C₁₀H₉NO₃
Molecular Weight: 191,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800435
CAS-Nummer: 127510-95-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR