7-Bromo-5-chloro-1,2-benzoxazole

Lieferant: Apollo Scientific

OR305029-250MG
APOSOR305029-250MGEA 66.7 EUR
APOSOR305029-250MG
7-Bromo-5-chloro-1,2-benzoxazole
7-Bromo-5-chloro-1,2-benzoxazole
Formel: C₇H₃BrClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD11847343
CAS-Nummer: 1260903-12-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR