1,3-Benzoxazole-4-carboxamide

Lieferant: Apollo Scientific

OR305030-250MG
APOSOR305030-250MGEA 509 EUR
APOSOR305030-250MG
1,3-Benzoxazole-4-carboxamide
1,3-Benzoxazole-4-carboxamide
Formel: C₈H₆N₂O₂
Molecular Weight: 162,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13176731
CAS-Nummer: 957995-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR