Benzo[d]oxazole-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR305031-1G
APOSOR305031-1GEA 358 EUR
APOSOR305031-1G
Benzo[d]oxazole-4-carboxylic acid
Benzo[d]oxazole-4-carboxylic acid
Formel: C₈H₅NO₃
Molecular Weight: 163,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11890806
CAS-Nummer: 208772-23-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR