Methyl 1,3-benzoxazole-4-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl benzo[d]oxazole-4-carboxylate

OR305032-250MG
APOSOR305032-250MGEA 88.6 EUR
APOSOR305032-250MG
Methyl 1,3-benzoxazole-4-carboxylate
Methyl 1,3-benzoxazole-4-carboxylate
Formel: C₉H₇NO₃
Molecular Weight: 177,157 g/mol
Schmelzpunkt: 107…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11110250
CAS-Nummer: 128156-54-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR