1,3-Benzothiazole-5-carbonitrile

Lieferant: Apollo Scientific

Synonyme: Benzo[d]thiazole-5-carbonitrile

OR305039-250MG
APOSOR305039-250MGEA 92.8 EUR
APOSOR305039-250MG
1,3-Benzothiazole-5-carbonitrile
1,3-Benzothiazole-5-carbonitrile
Formel: C₈H₄N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD06659663
CAS-Nummer: 58249-57-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR